Learn More
TARGETMOL CHEMICALS INC LATHOSTEROL 5MG
SDP
Catalog No. 502577350
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Lathosterol is a cholesterol-like endogenous metabolite. Serum Lathosterol concentration can be used as an indicator of whole-body cholesterol synthesis. purity: 99%
Catalog No. 50-257-7350 Supplier TARGETMOL CHEMICALS INC Supplier No. T157205MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Lathosterol is a cholesterol-like endogenous metabolite. Serum Lathosterol concentration can be used as an indicator of whole-body cholesterol synthesis. purity: 99%