Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lathyrol | 34420-19-4 | 334.45 | 50 MG
SDP

Catalog No. 5000388756
Encompass_Preferred

Lathyrol is a natural product identified for laboratory chemical uses and the manufacture of substances. It is utilized in cancer treatment, exhibiting reversal effects in MCF-7/ADR breast cancer cell lines in vitro. This product is intended for research use only and not for patient use.

  • Reverses effects in MCF-7/ADR breast cancer cell lines.
  • Intended for research use only.
  • Appears as a solid.
  • Color is white to off-white.
  • Molecular formula is C20H30O4.
  • Molecular weight is 334.45.
  • Purity is 99.89%.

Catalog No. 50-003-88756 Supplier Medchemexpress LLC Supplier No. HYN056150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lathyrol is a natural product identified for laboratory chemical uses and the manufacture of substances. It is utilized in cancer treatment, exhibiting reversal effects in MCF-7/ADR breast cancer cell lines in vitro. This product is intended for research use only and not for patient use.

  • Reverses effects in MCF-7/ADR breast cancer cell lines.
  • Intended for research use only.
  • Appears as a solid.
  • Color is white to off-white.
  • Molecular formula is C20H30O4.
  • Molecular weight is 334.45.
  • Purity is 99.89%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.