Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ Latitude™ HT Precast Gels

Catalog No. BMA57246 Shop All Lonza, Inc Products
Change view
Click to view available options
Wells:
2 x 50-well
4 x 25-well
Volume Per Well:
10 to 12 μL
25 to 30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wells Volume Per Well
BMA57246 4 x 25-well 25 to 30 μL
BMA57226 2 x 50-well 10 to 12 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Ethidium bromide is a powerful mutagen; gloves should be worn when opening pouches and handling gels and any solutions containing ethidium bromide
Catalog No. BMA57246 Supplier Lonza™ Supplier No. 57246
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethidium bromide is a powerful mutagen; gloves should be worn when opening pouches and handling gels and any solutions containing ethidium bromide

Ready-to-use gels for high-throughput DNA electrophoresis. Latitude™ HT Precast Gels are precision cast for accurate, reproducible resolution of DNA fragments for 8bp to 10kb with our SeaKem™ or NuSieve™ Agarose.

  • Provide rapid, reproducible DNA resolution of 8 bp to >=10kb
  • Manufactured from high-quality molecular biology grade SeaKem™ and NuSieve™ Brand Agarose
  • Quality and performance tested
  • Fit most large submerged electrophoresis chambers
  • Compatible with 8- and 12-tip multichannel pipets and microtiter plate spacing
  • Precision cast in accordance with ISO 9001 standards
  • Come in 100- and 200-well formats
  • Prestained with 0.5μg/mL of ethidium bromide
  • Permit runs to be completed in 30 minutes
  • Allow up to 200 samples to be run on a single precast gel

†Use of the reusable TruBand Anchor (sold separately) is required to ensure optimal gel performance

TRUSTED_SUSTAINABILITY

Specifications

Buffer TBE
Concentration 2%
Format Precast Gel
Separation Range 100 to ≥2000 bp
Wells 4 x 25-well
Dnase Free Yes
Formulation 2% SeaKem™ LE Plus Agarose, 1X TBE, Prestained with 0.5μg/mL of Ethidium Bromide
For Use With (Application) High throughput DNA electrophoresis
Material Agarose
Quantity 5 Gels
RNase Free Yes
Storage Requirements 18° to 26°C
Thickness (English) 0.25 in.
Thickness (Metric) 0.65 cm
Volume Per Well 25 to 30 μL
Length (Metric) Gel 24 cm
Width (Metric) Gel 14 cm
Product Type Agarose Gel
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.