Learn More
Cayman Chemical Latnoprost ethyl amIde-d4 50ug
SDP
Catalog No. 502991047
Encompass_Preferred
An internal standard for the quantification of latanoprost ethyl amide by GC- or LC-MS.
Catalog No. 50-299-1047 Supplier Cayman Chemical Supplier No. 31682250 UG
An internal standard for the quantification of latanoprost ethyl amide by GC- or LC-MS.