Learn More
Cayman Chemical Latnoprost ethyl amIded4 100ug
SDP
Catalog No. 502991360
Encompass_Preferred
An internal standard for the quantification of latanoprost ethyl amide by GC- or LC-MS.
Catalog No. 50-299-1360 Supplier Cayman Chemical Supplier No. 316822100 UG
An internal standard for the quantification of latanoprost ethyl amide by GC- or LC-MS.