Learn More
Cayman Chemical Latnoprost Lactol 10mg
SDP
Catalog No. 502896923
Encompass_Preferred
Latanoprost is an F-series prostaglandin analog which has been approved for use as an ocular hypotensive drug. Latanoprost lactol is an intermediate in the synthesis of latanoprost. Latanoprost lactol can be converted to the free acid of latanoprost by Wittig reaction with commercially available reagents.
Catalog No. 50-289-6923 Supplier Cayman Chemical Supplier No. 7004910 MG
Latanoprost is an F-series prostaglandin analog which has been approved for use as an ocular hypotensive drug. Latanoprost lactol is an intermediate in the synthesis of latanoprost. Latanoprost lactol can be converted to the free acid of latanoprost by Wittig reaction with commercially available reagents.