Learn More
Cayman Chemical Latanoprost Lactol, Purity: ≥98%, CAS: 352276-28-9, 50 mg
SDP
Catalog No. 502896898
Encompass_Preferred
Latanoprost is an F-series prostaglandin analog which has been approved for use as an ocular hypotensive drug. Latanoprost lactol is an intermediate in the synthesis of latanoprost. Latanoprost lactol can be converted to the free acid of latanoprost by Wittig reaction with commercially available reagents.
Catalog No. 50-289-6898 Supplier Cayman Chemical Supplier No. 7004950 MG
Latanoprost is an F-series prostaglandin analog which has been approved for use as an ocular hypotensive drug. Latanoprost lactol is an intermediate in the synthesis of latanoprost. Latanoprost lactol can be converted to the free acid of latanoprost by Wittig reaction with commercially available reagents.