Learn More
Cayman Chemical Latnoprost Lacton DIol 100mg
SDP
Catalog No. 502896710
Encompass_Preferred
Latanoprost is an F-series prostaglandin analog which has been approved for use as an ocular hypotensive drug. Latanoprost lactone diol is an intermediate in the synthesis of latanoprost. Latanoprost lactone diol can be converted to the free acid of latanoprost by reduction with DIBAL followed by Wittig reaction with commercially available reagents.
Catalog No. 50-289-6710 Supplier Cayman Chemical Supplier No. 70039100 MG
Latanoprost is an F-series prostaglandin analog which has been approved for use as an ocular hypotensive drug. Latanoprost lactone diol is an intermediate in the synthesis of latanoprost. Latanoprost lactone diol can be converted to the free acid of latanoprost by reduction with DIBAL followed by Wittig reaction with commercially available reagents.