Learn More
Cayman Chemical Latanoprost Lactone Diol, Purity: ≥98%, CAS: 145667-75-0, 50 mg
SDP
Catalog No. 502893579
Encompass_Preferred
Latanoprost is an F-series prostaglandin analog which has been approved for use as an ocular hypotensive drug. Latanoprost lactone diol is an intermediate in the synthesis of latanoprost. Latanoprost lactone diol can be converted to the free acid of latanoprost by reduction with DIBAL followed by Wittig reaction with commercially available reagents.
Catalog No. 50-289-3579 Supplier Cayman Chemical Supplier No. 7003950 MG
Latanoprost is an F-series prostaglandin analog which has been approved for use as an ocular hypotensive drug. Latanoprost lactone diol is an intermediate in the synthesis of latanoprost. Latanoprost lactone diol can be converted to the free acid of latanoprost by reduction with DIBAL followed by Wittig reaction with commercially available reagents.