Learn More
Cayman Chemical Latanoprost (free acid), Purity: ≥98%, CAS: 41639-83-2, 50 mg
SDP
Catalog No. 502965186
Encompass_Preferred
An FP receptor agonist and active metabolite of latanoprost; selectively binds to the FP receptor (Ki = 0.098 µM) over the EP1, EP2, EP3, and EP4 receptors (Kis = 2.06, 39.667, 7.519, and 75 µM, respectively), as well as the DP, IP, and TP receptors (Kis = ≥20, ≥90, and ≥60 µM, respectively); induces phosphoinositide turnover in isolated human ciliary muscle and human trabecular meshwork cells, mouse NIH3T3 fibroblasts, and rat A7r5 vascular smooth muscle cells, which all endogenously express FP receptors (EC50s = 124, 35, 32, and 35 nM, respectively), as well as HEK293 cells expressing human ocular FP receptors (EC50 = 45.7 nM); increases intracellular calcium levels in hEP1-5/293-AEQ17 cells expressing EP1 receptors in an aequorin-based calcium assay (EC50 = 119 nM); induces contraction of isolated cat iris sphincter smooth muscle (EC50 = 29.9 nM)
Catalog No. 50-296-5186 Supplier Cayman Chemical Supplier No. 1681150 MG
An FP receptor agonist and active metabolite of latanoprost; selectively binds to the FP receptor (Ki = 0.098 µM) over the EP1, EP2, EP3, and EP4 receptors (Kis = 2.06, 39.667, 7.519, and 75 µM, respectively), as well as the DP, IP, and TP receptors (Kis = ≥20, ≥90, and ≥60 µM, respectively); induces phosphoinositide turnover in isolated human ciliary muscle and human trabecular meshwork cells, mouse NIH3T3 fibroblasts, and rat A7r5 vascular smooth muscle cells, which all endogenously express FP receptors (EC50s = 124, 35, 32, and 35 nM, respectively), as well as HEK293 cells expressing human ocular FP receptors (EC50 = 45.7 nM); increases intracellular calcium levels in hEP1-5/293-AEQ17 cells expressing EP1 receptors in an aequorin-based calcium assay (EC50 = 119 nM); induces contraction of isolated cat iris sphincter smooth muscle (EC50 = 29.9 nM)