Learn More
Cayman Chemical Latnoprostfree acId-d4 50ug
SDP
Catalog No. 502991045
Encompass_Preferred
An internal standard for the quantification of latanoprost (free acid) by GC- or LC-MS
Catalog No. 50-299-1045 Supplier Cayman Chemical Supplier No. 31681150 UG
An internal standard for the quantification of latanoprost (free acid) by GC- or LC-MS