Learn More
Apexbio Technology LLC Latrepirdine 97657-92-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062343
Encompass_Preferred
Latrepirdine (CAS 97657-92-6) is an orally bioavailable small molecule investigated for its neuroprotective properties in preclinical studies of neurodegenerative diseases including Alzheimer s and Huntington s diseases Although its precise mechanism remains under investigation latrepirdine has demonstrated the ability to inhibit neuronal cell death pathways in animal disease models Moreover preliminary evidence indicates potential cognitive enhancement in normal physiological conditions Latrepirdine serves as a relevant tool compound to study neuronal survival mechanisms and cognitive function regulation
Catalog No. 50-000-62343 Supplier Apexbio Technology LLC Supplier No. B15725.1
Latrepirdine (CAS 97657-92-6) is an orally bioavailable small molecule investigated for its neuroprotective properties in preclinical studies of neurodegenerative diseases including Alzheimer s and Huntington s diseases Although its precise mechanism remains under investigation latrepirdine has demonstrated the ability to inhibit neuronal cell death pathways in animal disease models Moreover preliminary evidence indicates potential cognitive enhancement in normal physiological conditions Latrepirdine serves as a relevant tool compound to study neuronal survival mechanisms and cognitive function regulation