Learn More
Cayman Chemical Latrunculin A, 76343-93-6, 100 µg
SDP
Supplier:  Cayman Chemical 10010630100
Encompass_Preferred
A bioactive macrolide derived from sponges that binds monomeric actin, preventing actin polymerization both in vitro (Kd = 0.2 μM) and in cells (0.5 μM)
Catalog No. NC0673768