Learn More
FUJIFILM Biosciences Latrunculin A
Catalog No. 5000450307
Encompass_Preferred
Latrunculin A is an actin polymerization inhibitor It binds to actin in a 1 1 ratio and strongly inhibits its polymerization and has been reported as an inhibitor of fertilization division and early development in sea urchins and mice For research use only RUO
Catalog No. 50-004-50307 Supplier FUJIFILM Biosciences Supplier No. 12904361
Latrunculin A is an actin polymerization inhibitor It binds to actin in a 1 1 ratio and strongly inhibits its polymerization and has been reported as an inhibitor of fertilization division and early development in sea urchins and mice For research use only RUO