Learn More
Enzo Life Sciences Latrunculin A (500 µg). CAS: 76343-93-6
SDP
Supplier:  Enzo Life Sciences BMLT1190500
Encompass_Preferred
Inhibits actin polymerization, microfilament organzation and microfilament-mediated processes. Acts via a different mechanism and 10- to 20-fold more potently than cytochalasins. Inhibits phagocytosis by macrophages. Exposure to 0.2 mg/ml for one hour produces maximal change in cell morphology. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-2212