Learn More
Cayman Chemical Latrunculin A Inhibitors
SDP
Catalog No. 502887446
Encompass_Preferred
An actin disruption agent; sequesters G-actin and prevents F-actin assembly; binds monomeric actin with 1:1 stoichiometry and can be used to block actin polymerization both in vitro (Kd = 0.2 μM) and in cells (0.5 μM, 30 min); causes depolymerization of tumor cell cytoskeleton within ten minutes at 1-10 μM; suppresses actin synthesis at 10 μM; blocks dexamethasone-induced changes in actin cytoskeleton
Catalog No. 50-288-7446 Supplier Cayman Chemical Supplier No. 1001063025 UG
An actin disruption agent; sequesters G-actin and prevents F-actin assembly; binds monomeric actin with 1:1 stoichiometry and can be used to block actin polymerization both in vitro (Kd = 0.2 μM) and in cells (0.5 μM, 30 min); causes depolymerization of tumor cell cytoskeleton within ten minutes at 1-10 μM; suppresses actin synthesis at 10 μM; blocks dexamethasone-induced changes in actin cytoskeleton