Learn More
Abcam Latrunculin A (LAT-A), Actin polymerization inhibitor, 100UG
Catalog No. 502583282
Encompass_Preferred
MW 421.6 Da, Purity >95%. Actin polymerization inhibitor. Potent antiproliferative agent. Stabilizes monomeric G-actin. Shows greater potency than Latrunculin B (ab144291). Binds actin monomers near the nucleotide binding cleft with 1:1 stoichiometry. Inhibits growth of cancer cell lines (IC₅₀ values are 142, 142, 142, 166 and 95 nM for A549, H522-T1, HT-29, U-937 and MDA-MB-43 cells respectively). Shows convulsant effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3282 Supplier Abcam Supplier No. AB144290100UG
MW 421.6 Da, Purity >95%. Actin polymerization inhibitor. Potent antiproliferative agent. Stabilizes monomeric G-actin. Shows greater potency than Latrunculin B (ab144291). Binds actin monomers near the nucleotide binding cleft with 1:1 stoichiometry. Inhibits growth of cancer cell lines (IC₅₀ values are 142, 142, 142, 166 and 95 nM for A549, H522-T1, HT-29, U-937 and MDA-MB-43 cells respectively). Shows convulsant effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement