Learn More
Enzo Life Sciences Latrunculin A, (100µg), CAS Number: 76343-93-6
SDP
Supplier:  Enzo Life Sciences BMLT1190100
Encompass_Preferred
Ultra-pure; inhibits actin polymerization. Inhibits actin polymerization, microfilament organzation and microfilament-mediated processes. Acts via a different mechanism and 10- to 20-fold more potently than cytochalasins. Inhibits phagocytosis by macrophages. Exposure to 0.2 mg/ml for one hour produces maximal change in cell morphology. Formula: C22H31NO5S. MW: 421.6. CAS Number: 76343-93-6. MI: 14: 5378. Purity: ≥95% (HPLC). Appearance: White to yellow waxy solid. Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml). Use/Stability: Stable for 1 year after receipt when stored at -20°C. Stock solutions are stable for up to 3 months when stored at -20°C. Long Term Storage: -20°C
Catalog No. 50-103-1800