Learn More
AdipoGen Latrunculin B,Chemical. CAS: 76343-94-7
SDP
Supplier:  AdipoGen AGCN20031M001
Encompass_Preferred
Latrunculin B, AdipoGen Life Sciences. Chemical. CAS: 76343-94-7. Formula: C20H29NO5S. MW: 395.5. Isolated from Latrunculia magnifica. Cell permeable marine toxin. Disrupts microfilament-mediated processes. Actin polymerization inhibitor in vitro and in vivo by the formation of a 1:1 complex with monomeric G-actin. Depolymerizes actin filaments (F-actin). Less potent than Latrunculin A (Prod. No. AG-CN2-0027 http://www.adipogen.com/ag-cn2-0027/latrunculin-a.html ). May have fewer unwanted effects than latrunculin A and may be preferred for short-term studies. Acts via a different mechanism than cytochalasins.
Catalog No. 50-464-356