Learn More
Carnabio Usa Inc LATS1/10UG/CATALYTIC
Supplier:  Carnabio Usa Inc 0152310
Encompass
LATS1, Catalytic domain, Wild type, Amino Acid 589-1130, NP_004681.1, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0839