Learn More
Eurofins Discoverx LATS1, active, 10 µg. (14-988)
Supplier:  Eurofins Discoverx 14988
Encompass
N-terminal GST-tagged recombinant, human LATS1 amino acids 589-end and N-terminal 6xHis-tagged recombinant, human MOBKL1A full length. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9510