Learn More
Aviva Systems Biology Corp LATS1 Antibody - middle region (ARP58296_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58296_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LATS1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LATS1. Alternate names: WARTS, wtsApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-014-62279