Learn More
Eurofins Discoverx LATS2, active, 10 µg. (14-987)
Supplier:  Eurofins Discoverx 14987
Encompass
N-terminal GST-tagged recombinant, human LATS2 amino acids 553-end and N-terminal 6xHis-tagged recombinant, human MOBKL1A full length. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9508