Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bal Supply Lattice Connector
SCI_ED

Catalog No. S81082 Shop All Tip Industries Inc. Products
Change view
Click to view available options
Color:
Silver
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S81082 Silver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S81082 Supplier Bal Supply Supplier No. 4010LC/EMD

missing translation for 'validMainframeMsg'

Lattice Connector constructed of cast aluminum with a smooth round surface for connecting lattice rod supports for laboratory apparatus.

  • Rods can be joined at right angles in the same plan
  • Allows removal or addition of rods to a laboratory setup without disturbing the whole structure
  • Screws are easily tightened with the allen wrench
  • Flush screws eliminate the inconvenience of protruding locking devices

Specifications

Includes Wrench
Autoclavable Autoclavable
Color Silver
Holds Rods (0.5 in.)
Material Aluminum
Sterility Non-sterile
For Use With (Equipment) Lab Apparatus
Quantity 12/Pk
Size 0.5 in.
Weight (Metric) 0.5 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.