Learn More
Chemglass Life Sciences Chemglass Lattice Support Shelf, 9 in x 10 in
SDP
Catalog No. NC2457424
Encompass_Preferred
The Lattice Support Shelf is an anodized metal shelf that mounts to lattice support systems to provide a stable, slip-resistant work surface for laboratory items. It is designed for easy attachment and to prevent rotation once installed.
  • Anodized green finish.
  • Neoprene surface reduces slipping of items.
  • 1/2 in square support rod, 7.5 in long, prevents shelf rotation when mounted.
  • Attaches to lattice using the supplied clamp holder.
  • Part of the machined product line.
Catalog No. NC2457424 Supplier Chemglass Life Sciences Supplier No. CG305010SHELF
The Lattice Support Shelf is an anodized metal shelf that mounts to lattice support systems to provide a stable, slip-resistant work surface for laboratory items. It is designed for easy attachment and to prevent rotation once installed.
  • Anodized green finish.
  • Neoprene surface reduces slipping of items.
  • 1/2 in square support rod, 7.5 in long, prevents shelf rotation when mounted.
  • Attaches to lattice using the supplied clamp holder.
  • Part of the machined product line.