Learn More
Lauda Brinkmann Lp LAUDA Universa PRO
Supplier: Lauda Brinkmann Lp L003944
LAUDA Universa PROImmersion thermostats 100-125 V 50/60 HzTechnical FeaturesWorking temperature range 30 200 CWorking temperature range with external cooling 20 200 COperating temperature range -30 200 CAmbient temperature range 5 40 CTemperature stability 0 02 KHeating capacity range 1 1 1 5 kWCurrent max 12 APump Pressure max 0 6 barPump flow rate max (pressure) 22 L/minIn / Outlet connection thread (outside) M16 x 1Overall dimensions (WxDxH) 195 x 234 x 333 mmWeight 6 kgPower supply 100-125 V 50/60 HzPower plug Power cord with plug (NEMA 5-15P)