Learn More
Lauda Brinkmann Lp LAUDA Universa U 12 M
Catalog No. 5000573917
Encompass_Preferred
LAUDA Universa U 12 MHeating thermostat 200-240 V 50/60 HzTechnical FeaturesWorking temperature range 70 300 CWorking temperature range with external cooling 20 300 COperating temperature range -30 300 CAmbient temperature range 5 40 CTemperature stability 0 01 KHeating capacity range 2 8 3 7 kWCurrent max 16 ASize of bath (W x D x H) 200 x 200 x 200 mm Bath opening (WxT) 200 x 200 mmBath volume min / max 8 5 / 13 0 LPump Pressure max 0 7 barPump suction max 0 4 barPump flow rate max (pressure) 25 L/minPump Flow rate max (suction) 23 L/minIn / Outlet connection thread (outside) M16 x 1In / Outlet olives 13 mmOverall dimensions (WxDxH) 280 x 450 x 497 mmWeight 19 kgPower supply 200-240 V 50/60 HzPower plug Power cord with plug (NEMA 6-20P)
Catalog No. 50-005-73917 Supplier Lauda Brinkmann Lp Supplier No. L003777
LAUDA Universa U 12 MHeating thermostat 200-240 V 50/60 HzTechnical FeaturesWorking temperature range 70 300 CWorking temperature range with external cooling 20 300 COperating temperature range -30 300 CAmbient temperature range 5 40 CTemperature stability 0 01 KHeating capacity range 2 8 3 7 kWCurrent max 16 ASize of bath (W x D x H) 200 x 200 x 200 mm Bath opening (WxT) 200 x 200 mmBath volume min / max 8 5 / 13 0 LPump Pressure max 0 7 barPump suction max 0 4 barPump flow rate max (pressure) 25 L/minPump Flow rate max (suction) 23 L/minIn / Outlet connection thread (outside) M16 x 1In / Outlet olives 13 mmOverall dimensions (WxDxH) 280 x 450 x 497 mmWeight 19 kgPower supply 200-240 V 50/60 HzPower plug Power cord with plug (NEMA 6-20P)