Learn More
Lauda Brinkmann Lp LAUDA Universa U 16 M
Supplier: Lauda Brinkmann Lp L003879
LAUDA Universa U 16 MHeating thermostat 100-125 V 50/60 HzTechnical FeaturesWorking temperature range 70 300 CWorking temperature range with external cooling 20 300 COperating temperature range -30 300 CAmbient temperature range 5 40 CTemperature stability 0 01 KHeating capacity range 1 4 2 0 kWCurrent max 16 ASize of bath (W x D x H) 200 x 300 x 200 mm Bath opening (WxT) 200 x 300 mmBath volume min / max 11 5 / 17 0 LPump Pressure max 0 7 barPump suction max 0 4 barPump flow rate max (pressure) 25 L/minPump Flow rate max (suction) 23 L/minIn / Outlet connection thread (outside) M16 x 1In / Outlet olives 13 mmOverall dimensions (WxDxH) 280 x 550 x 497 mmWeight 22 kgPower supply 100-125 V 50/60 HzPower plug Power cord with plug (NEMA 5-20P)