Learn More
Lauda Brinkmann Lp LAUDA Universa U 16 P
Supplier: Lauda Brinkmann Lp L004238
LAUDA Universa U 16 PHeating thermostat 200-240 V 50/60 HzTechnical FeaturesWorking temperature range 40 200 CWorking temperature range with external cooling 20 200 COperating temperature range -30 200 CAmbient temperature range 5 40 CTemperature stability 0 01 KHeating capacity range 1 9 2 8 kWCurrent max 12 ASize of bath (W x D x H) 200 x 300 x 200 mm Bath opening (WxT) 200 x 300 mmBath volume min / max 11 5 / 17 0 LPump Pressure max 0 6 barPump flow rate max (pressure) 22 L/minIn / Outlet connection thread (outside) M16 x 1Overall dimensions (WxDxH) 280 x 550 x 476 mmWeight 18 kgPower supply 200-240 V 50/60 HzPower plug Power cord with plug (NEMA 6-20P)