Learn More
Lauda Brinkmann Lp LAUDA Universa U 1645 M
Catalog No. 5000573980
Encompass_Preferred
LAUDA Universa U 1645 MCooling thermostat 200-240 V 50/60 HzTechnical FeaturesWorking temperature range -45 200 COperating temperature range -45 200 CAmbient temperature range 5 40 CTemperature stability 0 01 KHeating capacity range 2 8 3 7 kWCurrent max 16 ASize of bath (W x D x H) 200 x 300 x 200 mm Bath opening (WxT) 200 x 300 mmBath volume min / max 10 5 / 16 5 LPump Pressure max 0 7 barPump suction max 0 4 barPump flow rate max (pressure) 25 L/minPump Flow rate max (suction) 23 L/minIn / Outlet connection thread (outside) M16 x 1In / Outlet olives 13 mmOverall dimensions (WxDxH) 310 x 610 x 757 mmWeight 49 kgRefrigerant stage 1 R-1270 (GWP 3) 0 075 kg 0 0 t CO2-eqPower supply 200-240 V 50/60 HzPower plug Power cord with plug (NEMA 6-20P)Cooling Capacity20 C (8) Ethanol 16 kW (50 Hz) 16 kW (60 Hz)-45 C (4) Ethanol 0 07 kW (50 Hz) 0 07 kW (60 Hz)
Catalog No. 50-005-73980 Supplier Lauda Brinkmann Lp Supplier No. L004163
LAUDA Universa U 1645 MCooling thermostat 200-240 V 50/60 HzTechnical FeaturesWorking temperature range -45 200 COperating temperature range -45 200 CAmbient temperature range 5 40 CTemperature stability 0 01 KHeating capacity range 2 8 3 7 kWCurrent max 16 ASize of bath (W x D x H) 200 x 300 x 200 mm Bath opening (WxT) 200 x 300 mmBath volume min / max 10 5 / 16 5 LPump Pressure max 0 7 barPump suction max 0 4 barPump flow rate max (pressure) 25 L/minPump Flow rate max (suction) 23 L/minIn / Outlet connection thread (outside) M16 x 1In / Outlet olives 13 mmOverall dimensions (WxDxH) 310 x 610 x 757 mmWeight 49 kgRefrigerant stage 1 R-1270 (GWP 3) 0 075 kg 0 0 t CO2-eqPower supply 200-240 V 50/60 HzPower plug Power cord with plug (NEMA 6-20P)Cooling Capacity20 C (8) Ethanol 16 kW (50 Hz) 16 kW (60 Hz)-45 C (4) Ethanol 0 07 kW (50 Hz) 0 07 kW (60 Hz)