Learn More
Lauda Brinkmann Lp LAUDA Universa U 20 M
Catalog No. 5000573976
Encompass_Preferred
LAUDA Universa U 20 MHeating thermostat 100-125 V 50/60 HzTechnical FeaturesWorking temperature range 65 300 CWorking temperature range with external cooling 20 300 COperating temperature range -30 300 CAmbient temperature range 5 40 CTemperature stability 0 01 KHeating capacity range 1 4 2 0 kWCurrent max 16 ASize of bath (W x D x H) 200 x 200 x 320 mm Bath opening (WxT) 200 x 200 mmBath volume min / max 9 5 / 22 0 LPump Pressure max 11 barPump flow rate max (pressure) 32 L/minIn / Outlet connection thread (outside) M16 x 1In / Outlet olives 13 mmOverall dimensions (WxDxH) 280 x 450 x 617 mmWeight 25 kgPower supply 100-125 V 50/60 HzPower plug Power cord with plug (NEMA 5-20P)
Catalog No. 50-005-73976 Supplier Lauda Brinkmann Lp Supplier No. L003863
LAUDA Universa U 20 MHeating thermostat 100-125 V 50/60 HzTechnical FeaturesWorking temperature range 65 300 CWorking temperature range with external cooling 20 300 COperating temperature range -30 300 CAmbient temperature range 5 40 CTemperature stability 0 01 KHeating capacity range 1 4 2 0 kWCurrent max 16 ASize of bath (W x D x H) 200 x 200 x 320 mm Bath opening (WxT) 200 x 200 mmBath volume min / max 9 5 / 22 0 LPump Pressure max 11 barPump flow rate max (pressure) 32 L/minIn / Outlet connection thread (outside) M16 x 1In / Outlet olives 13 mmOverall dimensions (WxDxH) 280 x 450 x 617 mmWeight 25 kgPower supply 100-125 V 50/60 HzPower plug Power cord with plug (NEMA 5-20P)