Learn More
Lauda Brinkmann Lp LAUDA Universa U 2040 M
Catalog No. 5000574027
Encompass_Preferred
LAUDA Universa U 2040 MCooling thermostat 200-240 V 50/60 HzTechnical FeaturesWorking temperature range -40 200 COperating temperature range -40 200 CAmbient temperature range 5 40 CTemperature stability 0 01 KHeating capacity range 2 8 3 7 kWCurrent max 16 ASize of bath (W x D x H) 200 x 200 x 320 mm Bath opening (WxT) 200 x 200 mmBath volume min / max 9 0 / 21 0 LPump Pressure max 11 barPump flow rate max (pressure) 32 L/minIn / Outlet connection thread (outside) M16 x 1In / Outlet olives 13 mmOverall dimensions (WxDxH) 350 x 540 x 927 mmWeight 57 kgRefrigerant stage 1 R-290 (GWP 3) 0 080 kg 0 0 t CO2-eqPower supply 200-240 V 50/60 HzPower plug Power cord with plug (NEMA 6-20P)Cooling Capacity20 C (8) Ethanol 0 8 kW (50 Hz) 0 8 kW (60 Hz)-40 C (4) Ethanol 0 1 kW (50 Hz) 0 1 kW (60 Hz)
Catalog No. 50-005-74027 Supplier Lauda Brinkmann Lp Supplier No. L003773
LAUDA Universa U 2040 MCooling thermostat 200-240 V 50/60 HzTechnical FeaturesWorking temperature range -40 200 COperating temperature range -40 200 CAmbient temperature range 5 40 CTemperature stability 0 01 KHeating capacity range 2 8 3 7 kWCurrent max 16 ASize of bath (W x D x H) 200 x 200 x 320 mm Bath opening (WxT) 200 x 200 mmBath volume min / max 9 0 / 21 0 LPump Pressure max 11 barPump flow rate max (pressure) 32 L/minIn / Outlet connection thread (outside) M16 x 1In / Outlet olives 13 mmOverall dimensions (WxDxH) 350 x 540 x 927 mmWeight 57 kgRefrigerant stage 1 R-290 (GWP 3) 0 080 kg 0 0 t CO2-eqPower supply 200-240 V 50/60 HzPower plug Power cord with plug (NEMA 6-20P)Cooling Capacity20 C (8) Ethanol 0 8 kW (50 Hz) 0 8 kW (60 Hz)-40 C (4) Ethanol 0 1 kW (50 Hz) 0 1 kW (60 Hz)