Learn More
Lauda Brinkmann Lp LAUDA Universa U 4 P
Catalog No. 5000573903
Encompass_Preferred
LAUDA Universa U 4 PHeating thermostat 200-240 V 50/60 HzTechnical FeaturesWorking temperature range 30 200 CWorking temperature range with external cooling 20 200 COperating temperature range -30 200 CAmbient temperature range 5 40 CTemperature stability 0 01 KHeating capacity range 1 9 2 8 kWCurrent max 12 ASize of bath (W x D x H) 130 x 100 x 160 mm Bath opening (WxT) 130 x 100 mmBath volume min / max 3 0 / 5 0 LPump Pressure max 0 6 barPump flow rate max (pressure) 22 L/minIn / Outlet connection thread (outside) M16 x 1Overall dimensions (WxDxH) 190 x 330 x 436 mmWeight 12 kgPower supply 200-240 V 50/60 HzPower plug Power cord with plug (NEMA 6-20P)
Catalog No. 50-005-73903 Supplier Lauda Brinkmann Lp Supplier No. L003906
LAUDA Universa U 4 PHeating thermostat 200-240 V 50/60 HzTechnical FeaturesWorking temperature range 30 200 CWorking temperature range with external cooling 20 200 COperating temperature range -30 200 CAmbient temperature range 5 40 CTemperature stability 0 01 KHeating capacity range 1 9 2 8 kWCurrent max 12 ASize of bath (W x D x H) 130 x 100 x 160 mm Bath opening (WxT) 130 x 100 mmBath volume min / max 3 0 / 5 0 LPump Pressure max 0 6 barPump flow rate max (pressure) 22 L/minIn / Outlet connection thread (outside) M16 x 1Overall dimensions (WxDxH) 190 x 330 x 436 mmWeight 12 kgPower supply 200-240 V 50/60 HzPower plug Power cord with plug (NEMA 6-20P)