Learn More
Lauda Brinkmann Lp LAUDA Universa U 40 P
Supplier: Lauda Brinkmann Lp L003909
LAUDA Universa U 40 PHeating thermostat 200-240 V 50/60 HzTechnical FeaturesWorking temperature range 40 200 CWorking temperature range with external cooling 20 200 COperating temperature range -30 200 CAmbient temperature range 5 40 CTemperature stability 0 01 KHeating capacity range 1 9 2 8 kWCurrent max 12 ASize of bath (W x D x H) 300 x 600 x 200 mm Bath opening (WxT) 300 x 600 mmBath volume min / max 27 5 / 41 0 LPump Pressure max 0 6 barPump flow rate max (pressure) 22 L/minIn / Outlet connection thread (outside) M16 x 1Overall dimensions (WxDxH) 380 x 850 x 478 mmWeight 29 kgPower supply 200-240 V 50/60 HzPower plug Power cord with plug (NEMA 6-20P)