Learn More
Lauda Brinkmann Lp LAUDA Universa U 420 P
Catalog No. 5000573926
Encompass_Preferred
LAUDA Universa U 420 PCooling thermostat 110-125 V 60 HzTechnical FeaturesWorking temperature range -20 200 COperating temperature range -20 200 CAmbient temperature range 5 40 CTemperature stability 0 02 KHeating capacity range 1 3 1 5 kWCurrent max 12 ASize of bath (W x D x H) 130 x 100 x 160 mm Bath opening (WxT) 130 x 100 mmBath volume min / max 1 8 / 4 0 LPump Pressure max 0 6 barPump flow rate max (pressure) 22 L/minIn / Outlet connection thread (outside) M16 x 1Overall dimensions (WxDxH) 210 x 410 x 616 mmWeight 26 kgRefrigerant stage 1 R-600a (GWP 3) 0 030 kg 0 0 t CO2-eqPower supply 110-127 V 60 HzPower plug Power cord with plug (NEMA 5-15P)Cooling Capacity20 C (6) Ethanol 0 2 kW (50 Hz) 0 2 kW (60 Hz)-20 C (3) Ethanol 0 07 kW (50 Hz) 0 07 kW (60 Hz)
Catalog No. 50-005-73926 Supplier Lauda Brinkmann Lp Supplier No. L003952
LAUDA Universa U 420 PCooling thermostat 110-125 V 60 HzTechnical FeaturesWorking temperature range -20 200 COperating temperature range -20 200 CAmbient temperature range 5 40 CTemperature stability 0 02 KHeating capacity range 1 3 1 5 kWCurrent max 12 ASize of bath (W x D x H) 130 x 100 x 160 mm Bath opening (WxT) 130 x 100 mmBath volume min / max 1 8 / 4 0 LPump Pressure max 0 6 barPump flow rate max (pressure) 22 L/minIn / Outlet connection thread (outside) M16 x 1Overall dimensions (WxDxH) 210 x 410 x 616 mmWeight 26 kgRefrigerant stage 1 R-600a (GWP 3) 0 030 kg 0 0 t CO2-eqPower supply 110-127 V 60 HzPower plug Power cord with plug (NEMA 5-15P)Cooling Capacity20 C (6) Ethanol 0 2 kW (50 Hz) 0 2 kW (60 Hz)-20 C (3) Ethanol 0 07 kW (50 Hz) 0 07 kW (60 Hz)