Learn More
Lauda Brinkmann Lp LAUDA Universa U 4230 M
Catalog No. 5000573934
Encompass_Preferred
LAUDA Universa U 4230 MCooling thermostat 100-125 V 50/60 HzTechnical FeaturesWorking temperature range -30 200 COperating temperature range -30 200 CAmbient temperature range 5 40 CTemperature stability 0 01 KHeating capacity range 1 4 2 0 kWCurrent max 16 ASize of bath (W x D x H) 300 x 350 x 320 mm Bath opening (WxT) 300 x 350 mmBath volume min / max 19 0 / 47 0 LPump Pressure max 11 barPump flow rate max (pressure) 32 L/minIn / Outlet connection thread (outside) M16 x 1In / Outlet olives 13 mmOverall dimensions (WxDxH) 450 x 690 x 927 mmWeight 69 kgRefrigerant stage 1 R-290 (GWP 3) 0 080 kg 0 0 t CO2-eqPower supply 100-125 V 50/60 HzPower plug Power cord with plug (NEMA 5-20P)Cooling Capacity20 C (8) Ethanol 0 8 kW (50 Hz) 0 8 kW (60 Hz)-30 C (4) Ethanol 0 18 kW (50 Hz) 0 18 kW (60 Hz)
Catalog No. 50-005-73934 Supplier Lauda Brinkmann Lp Supplier No. L003870
LAUDA Universa U 4230 MCooling thermostat 100-125 V 50/60 HzTechnical FeaturesWorking temperature range -30 200 COperating temperature range -30 200 CAmbient temperature range 5 40 CTemperature stability 0 01 KHeating capacity range 1 4 2 0 kWCurrent max 16 ASize of bath (W x D x H) 300 x 350 x 320 mm Bath opening (WxT) 300 x 350 mmBath volume min / max 19 0 / 47 0 LPump Pressure max 11 barPump flow rate max (pressure) 32 L/minIn / Outlet connection thread (outside) M16 x 1In / Outlet olives 13 mmOverall dimensions (WxDxH) 450 x 690 x 927 mmWeight 69 kgRefrigerant stage 1 R-290 (GWP 3) 0 080 kg 0 0 t CO2-eqPower supply 100-125 V 50/60 HzPower plug Power cord with plug (NEMA 5-20P)Cooling Capacity20 C (8) Ethanol 0 8 kW (50 Hz) 0 8 kW (60 Hz)-30 C (4) Ethanol 0 18 kW (50 Hz) 0 18 kW (60 Hz)