Learn More
Lauda Brinkmann Lp LAUDA Universa U 635 P
Supplier:  Lauda Brinkmann Lp L003915
Encompass_Preferred
LAUDA Universa U 635 PCooling thermostat 220-240 V 50/60 HzTechnical FeaturesWorking temperature range -35 200 COperating temperature range -35 200 CAmbient temperature range 5 40 CTemperature stability 0 02 KHeating capacity range 2 3 2 8 kWCurrent max 12 ASize of bath (W x D x H) 130 x 150 x 160 mm Bath opening (WxT) 130 x 150 mmBath volume min / max 3 2 / 5 7 LPump Pressure max 0 6 barPump flow rate max (pressure) 22 L/minIn / Outlet connection thread (outside) M16 x 1Overall dimensions (WxDxH) 290 x 480 x 646 mmWeight 34 kgRefrigerant stage 1 R-290 (GWP 3) 0 052 kg 0 0 t CO2-eqPower supply 220-240 V 50/60 HzPower plug Power cord with plug (NEMA 6-20P)Cooling Capacity20 C (6) Ethanol 0 5 kW (50 Hz) 0 5 kW (60 Hz)-35 C (3) Ethanol 0 02 kW (50 Hz) 0 02 kW (60 Hz)
Catalog No. 50-005-74039