Learn More
Lauda Brinkmann Lp LAUDA Universa U 8 P
Supplier:  Lauda Brinkmann Lp L003946
Encompass_Preferred
LAUDA Universa U 8 PHeating thermostat 100-125 V 50/60 HzTechnical FeaturesWorking temperature range 40 200 CWorking temperature range with external cooling 20 200 COperating temperature range -30 200 CAmbient temperature range 5 40 CTemperature stability 0 01 KHeating capacity range 1 1 1 5 kWCurrent max 12 ASize of bath (W x D x H) 150 x 150 x 200 mm Bath opening (WxT) 150 x 150 mmBath volume min / max 5 8 / 8 5 LPump Pressure max 0 6 barPump flow rate max (pressure) 22 L/minIn / Outlet connection thread (outside) M16 x 1Overall dimensions (WxDxH) 230 x 400 x 476 mmWeight 15 kgPower supply 100-125 V 50/60 HzPower plug Power cord with plug (NEMA 5-15P)
Catalog No. 50-005-74055