Learn More
Lauda Brinkmann Lp LAUDA Universa U 845 M
Catalog No. 5000573919
Encompass_Preferred
LAUDA Universa U 845 MCooling thermostat 100-125 V 50/60 HzTechnical FeaturesWorking temperature range -45 200 COperating temperature range -45 200 CAmbient temperature range 5 40 CTemperature stability 0 01 KHeating capacity range 1 4 2 0 kWCurrent max 16 ASize of bath (W x D x H) 150 x 150 x 200 mm Bath opening (WxT) 150 x 150 mmBath volume min / max 5 0 / 8 0 LPump Pressure max 0 7 barPump suction max 0 4 barPump flow rate max (pressure) 25 L/minPump Flow rate max (suction) 23 L/minIn / Outlet connection thread (outside) M16 x 1In / Outlet olives 13 mmOverall dimensions (WxDxH) 310 x 490 x 757 mmWeight 45 kgRefrigerant stage 1 R-290 (GWP 3) 0 080 kg 0 0 t CO2-eqPower supply 100-125 V 50/60 HzPower plug Power cord with plug (NEMA 5-20P)Cooling Capacity20 C (8) Ethanol 0 8 kW (50 Hz) 0 8 kW (60 Hz)-45 C (4) Ethanol 0 05 kW (50 Hz) 0 05 kW (60 Hz)
Catalog No. 50-005-73919 Supplier Lauda Brinkmann Lp Supplier No. L003865
LAUDA Universa U 845 MCooling thermostat 100-125 V 50/60 HzTechnical FeaturesWorking temperature range -45 200 COperating temperature range -45 200 CAmbient temperature range 5 40 CTemperature stability 0 01 KHeating capacity range 1 4 2 0 kWCurrent max 16 ASize of bath (W x D x H) 150 x 150 x 200 mm Bath opening (WxT) 150 x 150 mmBath volume min / max 5 0 / 8 0 LPump Pressure max 0 7 barPump suction max 0 4 barPump flow rate max (pressure) 25 L/minPump Flow rate max (suction) 23 L/minIn / Outlet connection thread (outside) M16 x 1In / Outlet olives 13 mmOverall dimensions (WxDxH) 310 x 490 x 757 mmWeight 45 kgRefrigerant stage 1 R-290 (GWP 3) 0 080 kg 0 0 t CO2-eqPower supply 100-125 V 50/60 HzPower plug Power cord with plug (NEMA 5-20P)Cooling Capacity20 C (8) Ethanol 0 8 kW (50 Hz) 0 8 kW (60 Hz)-45 C (4) Ethanol 0 05 kW (50 Hz) 0 05 kW (60 Hz)