Learn More
Lauda Brinkmann Lp LAUDA Universa U 845 P
Supplier:  Lauda Brinkmann Lp L004277
Encompass_Preferred
LAUDA Universa U 845 PCooling thermostat 200-240 V 50/60 HzTechnical FeaturesWorking temperature range -45 200 COperating temperature range -45 200 CAmbient temperature range 5 40 CTemperature stability 0 02 KHeating capacity range 1 9 2 8 kWCurrent max 12 ASize of bath (W x D x H) 150 x 150 x 200 mm Bath opening (WxT) 150 x 150 mmBath volume min / max 5 0 / 8 0 LPump Pressure max 0 6 barPump flow rate max (pressure) 22 L/minIn / Outlet connection thread (outside) M16 x 1Overall dimensions (WxDxH) 310 x 490 x 736 mmWeight 43 kgRefrigerant stage 1 R-290 (GWP 3) 0 080 kg 0 0 t CO2-eqPower supply 200-240 V 50/60 HzPower plug Power cord with plug (NEMA 6-20P)Cooling Capacity20 C (6) Ethanol 0 8 kW (50 Hz) 0 8 kW (60 Hz)-45 C (3) Ethanol 0 05 kW (50 Hz) 0 05 kW (60 Hz)
Catalog No. 50-005-73944