Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Tide Laundry Detergent, Refreshing Breeze, Liquid, 92 fl oz, 4/Case

Catalog No. 502867770
Encompass

This liquid laundry detergent is designed for effective cleaning in various laundry applications.

  • Scent: refreshing breeze
  • Form: liquid
  • Container size: 92 oz.
  • Formula type: detergent
  • Application: laundry
  • Package quantity: 4 bottles per case

Catalog No. 50-286-7770 Supplier MSC Supplier No. 20467387
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This liquid laundry detergent is designed for effective cleaning in various laundry applications.

  • Scent: refreshing breeze
  • Form: liquid
  • Container size: 92 oz.
  • Formula type: detergent
  • Application: laundry
  • Package quantity: 4 bottles per case

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.