Learn More
AdipoGen Laurdan,Chemical. CAS: 74515-25-6
SDP
Catalog No. 50596496
Encompass_Preferred
Laurdan, AdipoGen Life Sciences. Chemical. CAS: 74515-25-6. Formula: C24H35NO. MW: 353.54. Synthetic. N,N-Dimethyl-6-dodecanoyl-2-naphthylamine (Laurdan) is used as an environmentally sensitive probe for the labeling of plasma membranes. The lauroyl-sidechain integrates into the hydrophobic core of biological membranes, the polar fluorescent moiety is at level of the glycerol-backbone. Dipolar relaxation processes of the naphtylamine-moiety with neighboring polar molecules lead to wavelength-shifts of the emission-spectra. The physical state of the membrane and effects of temperature, cholesterol or ethanol on various membrane-systems have been monitored.
Catalog No. 50-596-496 Supplier AdipoGen Supplier No. CDXD0098M050
Laurdan, AdipoGen Life Sciences. Chemical. CAS: 74515-25-6. Formula: C24H35NO. MW: 353.54. Synthetic. N,N-Dimethyl-6-dodecanoyl-2-naphthylamine (Laurdan) is used as an environmentally sensitive probe for the labeling of plasma membranes. The lauroyl-sidechain integrates into the hydrophobic core of biological membranes, the polar fluorescent moiety is at level of the glycerol-backbone. Dipolar relaxation processes of the naphtylamine-moiety with neighboring polar molecules lead to wavelength-shifts of the emission-spectra. The physical state of the membrane and effects of temperature, cholesterol or ethanol on various membrane-systems have been monitored.