Learn More
AnaSpec LAURDAN
Catalog No. 50850456
Encompass
Laurdan[[6-Dodecanoyl-2-dimethylaminonaphthalene]] ; MW 353.6; (25 mg) Environment-sensitive dye for studying membrane and protein structures
Catalog No. 50-850-456 Supplier AnaSpec Supplier No. AS88201
Laurdan[[6-Dodecanoyl-2-dimethylaminonaphthalene]] ; MW 353.6; (25 mg) Environment-sensitive dye for studying membrane and protein structures