Learn More
Medchemexpress LLC Lauric acid-1-13C | 93639-08-8 | MFCD00002737 | >95.0% | 201.3 | C11[13C]H24O2 | 100mg
SDP
Catalog No. 5000268458
Encompass_Preferred
Lauric acid-13C is the 13C labeled Lauric acid Lauric acid is a middle chain-free fatty acid with strong bactericidal properties The EC50s for P acnes S aureus S epidermidis are 2 6 4 g/mL respectively
Catalog No. 50-002-68458 Supplier Medchemexpress LLC Supplier No. HYY0366S100MG
Lauric acid-13C is the 13C labeled Lauric acid Lauric acid is a middle chain-free fatty acid with strong bactericidal properties The EC50s for P acnes S aureus S epidermidis are 2 6 4 g/mL respectively