Learn More
Apexbio Technology LLC Lauric Acid 143-07-7 500mg
SDP
Catalog No. 5000090879
Encompass_Preferred
Lauric acid (143-07-7) is a small-molecule modulator designed to induce apoptosis and inhibit microbial colonization thereby regulating cell viability and inflammatory responses Lauric acid exerts its biological activity primarily through ROS-dependent oxidative stress cell cycle arrest at S and G2/M phases and reduction of intracellular glutathione In cell-based studies lauric acid induces apoptosis in colon carcinoma cells (Caco-2 and IEC-6) and reduces Propionibacterium acnes colonization in animal models Based on these pharmacological properties lauric acid holds research potential in investigating antiproliferative mechanisms in cancer and antimicrobial applications including acne treatment
Catalog No. 50-000-90879 Supplier Apexbio Technology LLC Supplier No. B6155500
Lauric acid (143-07-7) is a small-molecule modulator designed to induce apoptosis and inhibit microbial colonization thereby regulating cell viability and inflammatory responses Lauric acid exerts its biological activity primarily through ROS-dependent oxidative stress cell cycle arrest at S and G2/M phases and reduction of intracellular glutathione In cell-based studies lauric acid induces apoptosis in colon carcinoma cells (Caco-2 and IEC-6) and reduces Propionibacterium acnes colonization in animal models Based on these pharmacological properties lauric acid holds research potential in investigating antiproliferative mechanisms in cancer and antimicrobial applications including acne treatment