Learn More
Medchemexpress LLC Lauric acid | 143-07-7 | MFCD00002736 | 99.9% | 200.32 g/mol | C12H24O2 | 10g
SDP
Catalog No. 5000268363
Encompass_Preferred
Lauric acid is a middle chain-free fatty acid with strong bactericidal properties The EC50s for P acnes S aureus S epidermidis are 2 6 4 g/mL respectively[1]
Catalog No. 50-002-68363 Supplier Medchemexpress LLC Supplier No. HYY036610G
Lauric acid is a middle chain-free fatty acid with strong bactericidal properties The EC50s for P acnes S aureus S epidermidis are 2 6 4 g/mL respectively[1]