Learn More
Medchemexpress LLC HY-Y0366 1g Medchemexpress, Lauric acid CAS:143-07-7 Purity:>98%
SDP
Catalog No. 501934687
Encompass_Preferred
Medchemexpress, HY-Y0366 1g Lauric acid CAS:143-07-7 Lauric acid is a middle chain-free fatty acid with strong bactericidal properties. The EC 50 s for P. acnes, S.aureus, S. epidermidis, are 2, 6, 4 μg/mL, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4687 Supplier Medchemexpress LLC Supplier No. HYY03661G
Medchemexpress, HY-Y0366 1g Lauric acid CAS:143-07-7 Lauric acid is a middle chain-free fatty acid with strong bactericidal properties. The EC 50 s for P. acnes, S.aureus, S. epidermidis, are 2, 6, 4 μg/mL, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.