Learn More
Selleck Chemical LLC Lauric Acid S4726-500mg
SDP
Catalog No. 503111974
Encompass_Preferred
Lauric Acid (dodecanoic acid N-Dodecanoic acid Dodecylic acid) is a saturated medium-chain fatty acid with a 12-carbon backbone Lauric acid is found naturally in various plant and animal fats and oils and is a major component of coconut oil and palm kernel oil
Catalog No. 50-311-1974 Supplier Selleck Chemical LLC Supplier No. S4726500MG
Lauric Acid (dodecanoic acid N-Dodecanoic acid Dodecylic acid) is a saturated medium-chain fatty acid with a 12-carbon backbone Lauric acid is found naturally in various plant and animal fats and oils and is a major component of coconut oil and palm kernel oil