Learn More
Cayman Chemical Lauric Acid-d2, Purity: ≥99% deuterated forms (d1-d2), CAS: 64118-39-4, 250 mg
SDP
Catalog No. 502992383
Encompass_Preferred
An internal standard for the quantification of lauric acid by GC- or LC-MS
Catalog No. 50-299-2383 Supplier Cayman Chemical Supplier No. 29410250 MG
An internal standard for the quantification of lauric acid by GC- or LC-MS