Learn More
Cayman Chemical Lauric Acid-d2, Purity: ≥99% deuterated forms (d1-d2), CAS: 64118-39-4, 50 mg
SDP
Catalog No. 502990070
Encompass_Preferred
An internal standard for the quantification of lauric acid by GC- or LC-MS
Catalog No. 50-299-0070 Supplier Cayman Chemical Supplier No. 2941050 MG
An internal standard for the quantification of lauric acid by GC- or LC-MS